Cerium(III) carbonate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Cerium(III) carbonate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cerium(III) carbonate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 015295.22
also known as 015295-22
Price (USD)/ Each
78.65
Online exclusive
87.40 
Save 8.75 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
78.65
Online exclusive
87.40 
Save 8.75 (10%)
Add to cart
Cerium(III) carbonate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Catalog number015295.22
Price (USD)/ Each
78.65
Online exclusive
87.40 
Save 8.75 (10%)
-
Add to cart
Chemical Identifiers
CAS54451-25-1
IUPAC Namedicerium(3+) tricarbonate
Molecular FormulaC3Ce2O9
InChI KeyGHLITDDQOMIBFS-UHFFFAOYSA-H
SMILES[Ce+3].[Ce+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Raw material of various high purity cerium complexes and high grad polishing powder. Cerium Carbonate is used to decolorize glass by keeping iron in its ferrous state.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only