Thermo Scientific Chemicals

Sodium 1-pentanesulfonate monohydrate, HPLC grade, Thermo Scientific Chemicals

Catalog number: 015714.06
5 g, Each
Thermo Scientific Chemicals

Sodium 1-pentanesulfonate monohydrate, HPLC grade, Thermo Scientific Chemicals

Catalog number: 015714.06
5 g, Each
Quantity
Catalog number: 015714.06
also known as 015714-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
207605-40-1
IUPAC Name
pentane-1-sulfonate
Molecular Formula
C5H11O3S
InChI Key
RJQRCOMHVBLQIH-UHFFFAOYSA-M
SMILES
CCCCCS([O-])(=O)=O
Appearance (Color)
White
Assay (unspecified)
≥98.0%
Optical Absorbance at
210nm: <0.200 AU ( 0.25M aq. solution)
Optical Absorbance at
240nm: <0.050 AU ( 0.25M aq. solution)
Optical Absorbance at
250nm: <0.050 AU ( 0.25M aq. solution)

Description

Ion pair reagent.Sodium 1-pentanesulfonate monohydrate is used as a surfactant and a reversed-phase ion-pairing agent. It is utilized to study its effects on the properties of polyaniline, using Fourier transform infrared and UV-visible spectroscopy. Further, it is used for HPLC analysis of proteins and peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text