Sodium 1-octanesulfonate monohydrate, 99+%, Thermo Scientific Chemicals
Sodium 1-octanesulfonate monohydrate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium 1-octanesulfonate monohydrate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 015764.09
also known as 015764-09
Price (USD)/ Each
67.65
Online exclusive
75.00 
Save 7.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
67.65
Online exclusive
75.00 
Save 7.35 (10%)
Add to cart
Sodium 1-octanesulfonate monohydrate, 99+%, Thermo Scientific Chemicals
Catalog number015764.09
Price (USD)/ Each
67.65
Online exclusive
75.00 
Save 7.35 (10%)
-
Add to cart
Chemical Identifiers
CAS207596-29-0
IUPAC Namesodium hydrate octane-1-sulfonate
Molecular FormulaC8H19NaO4S
InChI KeyMBURIAHQXJQKRE-UHFFFAOYSA-M
SMILESO.[Na+].CCCCCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Assay (unspecified)≥99.0%
Loss on Drying7 to 9%
Optical Absorbance at200nm: ≤0.30AU
Optical Absorbance at210nm: ≤0.20AU
View more
Sodium 1-octanesulfonate monohydrate is used as an anionic detergent. It is an ion pairing reagent used in the HPLC analysis of proteins and peptides. Further, it is used as a surface active agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only