Thermo Scientific Chemicals

Oxalacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 015789.09
10 g, Each
Thermo Scientific Chemicals

Oxalacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 015789.09
10 g, Each
Quantity
Catalog number: 015789.09
also known as 015789-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
328-42-7
IUPAC Name
2-oxobutanedioic acid
Molecular Formula
C4H4O5
InChI Key
KHPXUQMNIQBQEV-UHFFFAOYSA-N
SMILES
OC(=O)CC(=O)C(O)=O
Appearance (Color)
White to off-white
Form
Powder or crystalline powder
Assay from Suppliers CofA
≥98.0 to ≤102.0% (Titration)
Chloride content
≤0.01%
Residue on ignition
≤0.1% (as sulfate)

Description

Oxaloacetic acid is a substrate for malate dehydrogenase and oxaloacetate decarboxylase. It is an inhibitor of succinic dehydrogenase. It is an intermediate in the citric acid cycle and glucogenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text