Thermo Scientific Chemicals
Perfluorohexanes, 98+%, Thermo Scientific Chemicals
Catalog number: 015812.14
25 g, Each
Thermo Scientific Chemicals
Perfluorohexanes, 98+%, Thermo Scientific Chemicals
Catalog number: 015812.14
25 g, Each
Quantity
Catalog number: 015812.14
Price (USD)
Have Questions?
Description
Perfluorohexanes is used as a coolant, photosensitizer in fluorous biphasic singlet oxygenation and an inert reaction medium. It is used in the electronic cooling liquid and insulator. It plays a vital role as a reaction medium, especially for photooxidation reactions. Further, it finds application as an ultrasound contrast agent.