Tetra-n-butylammonium hydroxide, 55% w/w aq. soln., Thermo Scientific Chemicals
Tetra-n-butylammonium hydroxide, 55% w/w aq. soln., Thermo Scientific Chemicals
Thermo Scientific™

Tetra-n-butylammonium hydroxide, 55% w/w aq. soln., Thermo Scientific Chemicals

 
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Catalog number 016198.30
also known as 016198-30
Price (USD)/ Each
205.65
Online exclusive
228.00 
Save 22.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
205.65
Online exclusive
228.00 
Save 22.35 (10%)
Add to cart
Tetra-n-butylammonium hydroxide, 55% w/w aq. soln., Thermo Scientific Chemicals
Catalog number016198.30
Price (USD)/ Each
205.65
Online exclusive
228.00 
Save 22.35 (10%)
-
Add to cart
Chemical Identifiers
CAS2052-49-5
IUPAC Nametetrabutylazanium hydroxide
Molecular FormulaC16H37NO
InChI KeyVDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES[OH-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Trace MetalSodium, Na + potassium, K: ≤0.2%
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (Titration)≥52.5%
Residual halide≤0.5% as bromide
View more

As a reagent for ion-pair extractionsTetra-n-butylammonium hydroxide, 55% w/w aq. solution is used as a phase-transfer catalyst in organic synthesis especially for alkylation and condensation reactions. It is used as a curing accelerator for epoxy resins. It is also used in preparation of dichlorocarbene from chloroform.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
RUO – Research Use Only