Thermo Scientific Chemicals

Thymolphthalein, ACS

Catalog number: 016245.06
5 g, Each
Thermo Scientific Chemicals

Thymolphthalein, ACS

Catalog number: 016245.06
5 g, Each
Quantity
Catalog number: 016245.06
also known as 016245-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
125-20-2
IUPAC Name
3,3-bis[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]-1,3-dihydro-2-benzofuran-1-one
Molecular Formula
C28H30O4
InChI Key
LDKDGDIWEUUXSH-UHFFFAOYSA-N
SMILES
CC(C)C1=CC(=C(C)C=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C=C1C
Comment
Clarity of solution P.T.
Comment
Visual transition interval from pH 8.8 (colorless) to 10.5 (blue)

Description

It acts as pH indicator, as reagent for blood after decolorizing the alkaline solution by boiling with zinc dust.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text