Tetra-n-butylammonium tribromide, 98%, Thermo Scientific Chemicals
Tetra-n-butylammonium tribromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetra-n-butylammonium tribromide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 016258.22
also known as 016258-22
Price (USD)/ Each
152.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
152.00
Add to cart
Tetra-n-butylammonium tribromide, 98%, Thermo Scientific Chemicals
Catalog number016258.22
Price (USD)/ Each
152.00
-
Add to cart
Chemical Identifiers
CAS38932-80-8
IUPAC NameN,N,N-trimethylanilinium
Molecular FormulaC9H14N
InChI KeyZNEOHLHCKGUAEB-UHFFFAOYSA-N
SMILESC[N+](C)(C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥97.5% (1H-NMR)
Proton NMRConforms to structure
FormCrystalline powder or powder
Melting Point (clear melt)71-75°C
Appearance (Color)Orange
Tetra-n-butylammonium tribromide is used as a phase transfer catalyst in organic synthesis as well as a mild brominating agent. It acts as a solid source of bromine. It is useful for the preparation of vicinal dibromides from alkenes and alkynes and alfa-bromo acetals. Further, it is utilized to generate hydrogen bromide, which is used as an efficient chemoselective reagent for acetalization of carbonyl compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only