Thermo Scientific Chemicals

alpha-Terpineol, 96%, Thermo Scientific Chemicals

Catalog number: 016285.36
500 g, Each
Thermo Scientific Chemicals

alpha-Terpineol, 96%, Thermo Scientific Chemicals

Catalog number: 016285.36
500 g, Each
Quantity
Catalog number: 016285.36
also known as 016285-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98-55-5
IUPAC Name
2-[(1R)-4-methylcyclohex-3-en-1-yl]propan-2-ol
Molecular Formula
C10H18O
InChI Key
WUOACPNHFRMFPN-VIFPVBQESA-N
SMILES
CC1=CC[C@@H](CC1)C(C)(C)O
Form
Liquid/solidified melt
Assay (GC)
95% min ( α + ϒ)
Comment
Product may congeal in cold or room temperature. Warm and mix thoroughly to homogenize liquid before using or sampling

Description

A naturally-occuring monoterpene alcohol.Alpha-terpineol is used as an antioxidant, antiseptic, antihypernociception and anti-inflammatory. It is also used as a solvent. It is an important ingredient of pine oil disinfectants. Further, it is used as a fragrance in perfumes, fat denaturant for soap production and synthetic flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text