Thermo Scientific Chemicals

Phenol Red, ACS

Catalog number: 016294.22
100 g, Each
Thermo Scientific Chemicals

Phenol Red, ACS

Catalog number: 016294.22
100 g, Each
Quantity
Catalog number: 016294.22
also known as 016294-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
143-74-8
IUPAC Name
3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular Formula
C19H14O5S
InChI Key
BELBBZDIHDAJOR-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Comment
Clarity of solution P.T.
Comment
Visual transition interval from pH 6.8 (yellow) to 8.2 (red)

Description

Phenol red is used as non absorbed marker in the gravimetric method. It is a pH indicator used in cell biology laboratories and in home swimming pool test kits. It is used to check the kidney function based on the excreted phenol red by colorimetric method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text