Thermo Scientific Chemicals

Pyrazine, 98%, Thermo Scientific Chemicals

Catalog number: 016326.09
10 g, Each
Thermo Scientific Chemicals

Pyrazine, 98%, Thermo Scientific Chemicals

Catalog number: 016326.09
10 g, Each
Quantity
Catalog number: 016326.09
also known as 016326-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
290-37-9
IUPAC Name
pyrazine
Molecular Formula
C4H4N2
InChI Key
KYQCOXFCLRTKLS-UHFFFAOYSA-N
SMILES
C1=CN=CC=N1
Form
Powder or crystals or crystalline powder or flakes
Appearance (Color)
White to off-white
Assay from Supplier's CofA
≥97.5%
Melting Point
51.0-56.0°

Description

Pyrazine is used as a flavoring agent. It serves as a component of some herbs in traditional Chinese medicine. It is employed as pharmaceutical intermediates, perfume intermediates and involved in the preparation of other chemicals. In addition to this, it is used in the production of pyrazine 1-oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text