Thermo Scientific Chemicals

Tetraethylammonium hydroxide, 25% w/w in methanol, Thermo Scientific Chemicals

Catalog number: 016328.18
50 g, Each
Thermo Scientific Chemicals

Tetraethylammonium hydroxide, 25% w/w in methanol, Thermo Scientific Chemicals

Catalog number: 016328.18
50 g, Each
Quantity
Catalog number: 016328.18
also known as 016328-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-98-5
IUPAC Name
tetraethylazanium hydroxide
Molecular Formula
C8H21NO
InChI Key
LRGJRHZIDJQFCL-UHFFFAOYSA-M
SMILES
[OH-].CC[N+](CC)(CC)CC
Assay from Supplier's CofA
≥24.0 to ≤26.0% (titration)
Proton NMR
Conforms
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid

Description

It is used as a phase transfer catalyst, the template agent synthesized by molecular sieve and cleaning agents, as a surfactant, used to anisotropically etch silicon, in thermochemolysis, as a basic solvent in the development of acidic photo resist in the photolithography process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text