Thermo Scientific Chemicals

Xylenes, ACS, 98.5+% (Assay, isomers plus ethylbenzene), Thermo Scientific Chemicals

Catalog number: 016371.K7
4 L, Each
Thermo Scientific Chemicals

Xylenes, ACS, 98.5+% (Assay, isomers plus ethylbenzene), Thermo Scientific Chemicals

Catalog number: 016371.K7
4 L, Each
Quantity
Catalog number: 016371.K7
also known as 016371-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1330-20-7
IUPAC Name
1,3-xylene
Molecular Formula
C8H10
InChI Key
IVSZLXZYQVIEFR-UHFFFAOYSA-N
SMILES
CC1=CC(C)=CC=C1
Form
Clear liquid
Impurity content
≤0.003% Sulfur compounds (as S)
Impurity content
Substances darkened by sulfuric acid P.T.
Residue on Evaporation
≤0.002%
Color (APHA)
≤10

Description

Xylene is used as a solvent in pharmaceutical, printing, rubber, and leather industries. It is an important component of ink, rubber and adhesives as well as in thinning paints and varnishes. It is used in the laboratory to make baths with dry ice to cool reaction vessels, and as a solvent to remove synthetic immersion oil from the microscope objective in light microscopy. It finds application as a short-term hydrophobic clearing agent of tissue samples by histologists. It is a precursor for the preparation of xylene dichlorides, xylyl bromide, terephthalic acid and dimethyl terephthalate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text