Cesium hydroxide, 50% w/w aq. soln., 99% (metals basis), Thermo Scientific Chemicals
Cesium hydroxide, 50% w/w aq. soln., 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cesium hydroxide, 50% w/w aq. soln., 99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 016505.MZ
also known as 016505-MZ
Price (USD)/ Each
830.65
Online exclusive
923.00 
Save 92.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
830.65
Online exclusive
923.00 
Save 92.35 (10%)
Add to cart
Cesium hydroxide, 50% w/w aq. soln., 99% (metals basis), Thermo Scientific Chemicals
Catalog number016505.MZ
Price (USD)/ Each
830.65
Online exclusive
923.00 
Save 92.35 (10%)
-
Add to cart
Chemical Identifiers
CAS21351-79-1
IUPAC Namecaesium(1+) hydroxide
Molecular FormulaCsHO
InChI KeyHUCVOHYBFXVBRW-UHFFFAOYSA-M
SMILES[OH-].[Cs+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Cesium hydroxide is used in the preparation of porous cesium titanosilicates, which are potential ion-exchange materials utilized for radioactive wastes cleaning purpose. It is also involved in the preparation of other cesium salts. Further, it is used as an electrolyte in alkaline batteries and as a catalyst in organic synthesis. In addition to this, it is used in color photography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only