Ruthenium(III) nitrosylchloride, Ru 42.1% min, Thermo Scientific Chemicals
Ruthenium(III) nitrosylchloride, Ru 42.1% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ruthenium(III) nitrosylchloride, Ru 42.1% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 016622.06
also known as 016622-06
Price (USD)/ Each
509.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
509.00
Add to cart
Ruthenium(III) nitrosylchloride, Ru 42.1% min, Thermo Scientific Chemicals
Catalog number016622.06
Price (USD)/ Each
509.00
-
Add to cart
Chemical Identifiers
CAS18902-42-6
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Ru 42.1% min
Ruthenium nitrosyl chloride is an important ruthenium compounds, which is used in synthesis of more complex ruthenium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only