Lanthanum(III) carbonate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Lanthanum(III) carbonate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lanthanum(III) carbonate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number 016639.22
also known as 016639-22
Price (USD)/ Each
57.60
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
57.60
Add to cart
Lanthanum(III) carbonate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Catalog number016639.22
Price (USD)/ Each
57.60
-
Add to cart
Chemical Identifiers
CAS54451-24-0
IUPAC Namedilanthanum(3+) tricarbonate
Molecular FormulaC3La2O9
InChI KeyNZPIUJUFIFZSPW-UHFFFAOYSA-H
SMILES[La+3].[La+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.

It is used in pharmaceuticals & used in medicine as a phosphate binder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only