Hydrazine monohydrate, 99+%, Thermo Scientific Chemicals
Hydrazine monohydrate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrazine monohydrate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 500 g
100 g
500 g
2 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 016651.57
also known as 016651-57
Price (USD)/ Each
398.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
469.00 
Save 70.35 (15%)
-
Add to cart
Quantity:
4 x 500 g
Request bulk or custom format
Price (USD)/ Each
398.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
469.00 
Save 70.35 (15%)
Add to cart
Hydrazine monohydrate, 99+%, Thermo Scientific Chemicals
Catalog number016651.57
Price (USD)/ Each
398.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
469.00 
Save 70.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7803-57-8
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)> 99.0%
Hydrazine Hydrate used as a reactant in the cyclizations of pyridinones. It is also used in the study of nanocrystal semiconductors, participating in the functionalization and passivation of surface states. It is widely used as a reducing agent or an intermediate of synthesis in various industrial sectors like water treatment (effluents, industrial boilers), chemical treatment process (metals, mine extraction) or active ingredients synthesis (pharmaceuticals and agrochemicals).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only