Thermo Scientific Chemicals

Strontium chloride, anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 016790.14
25 g, Each
Thermo Scientific Chemicals

Strontium chloride, anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 016790.14
25 g, Each
Quantity
Catalog number: 016790.14
also known as 016790-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10476-85-4
IUPAC Name
strontium(2+) hexahydrate dichloride
Molecular Formula
Cl2H12O6Sr
InChI Key
AMGRXJSJSONEEG-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
Appearance (Color)
White
Particle Size
-40 Mesh
Total Metal Impurities
≤0.5% (5000 ppm)
Form
Powder

Description

Strontium chloride is widely used in fireworks for the source of red color. It is also used in dental and biological research. It is the precursor material to other compounds of strontium. It is used to prepare yellow strontium chromate, a corrosion inhibitor for aluminum. It is also employed in glass-making, sea water aquaria, and metallurgy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text