Thermo Scientific Chemicals

4-Bromo-1-butanol, tech., cont. varying amounts of THF, Thermo Scientific Chemicals

Catalog number: 016805.14
25 g, Each
Thermo Scientific Chemicals

4-Bromo-1-butanol, tech., cont. varying amounts of THF, Thermo Scientific Chemicals

Catalog number: 016805.14
25 g, Each
Quantity
Catalog number: 016805.14
also known as 016805-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
33036-62-3
IUPAC Name
4-bromobutan-1-ol
Molecular Formula
C4H9BrO
InChI Key
SIJLYRDVTMMSIP-UHFFFAOYSA-N
SMILES
OCCCCBr
Assay (unspecified)
> 90.0%
Appearance (Color)
Clear colorless to pale yellow
Form
liquid

Description

4-Bromo-1-butanol is used to prepare 2-(4-bromo-butoxy)-tetrahydro-pyran by reacting with 3,4-dihydro-2H-pyran in the presence of amberlyst 15 as a catalyst. It reacts with dihydropyran to prepare tetrahydropyranyl ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text