Thermo Scientific Chemicals

Azomethine-H hydrate, Thermo Scientific Chemicals

Catalog number: 016810.04
2 g, Each
Thermo Scientific Chemicals

Azomethine-H hydrate, Thermo Scientific Chemicals

Catalog number: 016810.04
2 g, Each
Quantity
Catalog number: 016810.04
also known as 016810-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
32266-60-7
IUPAC Name
4-hydroxy-5-[(Z)-[(2-hydroxyphenyl)methylidene]amino]naphthalene-2,7-disulfonic acid
Molecular Formula
C17H13NO8S2
InChI Key
DUCCKQSNXPFEGT-NVMNQCDNSA-N
SMILES
OC1=CC=CC=C1\C=N/C1=C2C(O)=CC(=CC2=CC(=C1)S(O)(=O)=O)S(O)(=O)=O
Form
Powder
Proton NMR
Consistent with structure
Assay by NMR
>95%

Description

Azomethine-H hydrate is used as a highly sensitive and colorimetric reagent for testing and detection of boron. It is also used to detect microgram levels of boron in glass and steel samples. Further, it is involved in the detection of boron in plant samples by masking copper, iron and aluminum ions with ethylenediaminetetraacetic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text