Azomethine-H hydrate, Thermo Scientific Chemicals
Azomethine-H hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Azomethine-H hydrate, Thermo Scientific Chemicals

Reagent for boron determination
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
2 g
10 g
Catalog number 016810.14
also known as 016810-14
Price (USD)/ Each
444.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
444.00
Add to cart
Azomethine-H hydrate, Thermo Scientific Chemicals
Catalog number016810.14
Price (USD)/ Each
444.00
-
Add to cart
Chemical Identifiers
CAS32266-60-7
IUPAC Name4-hydroxy-5-[(Z)-[(2-hydroxyphenyl)methylidene]amino]naphthalene-2,7-disulfonic acid
Molecular FormulaC17H13NO8S2
InChI KeyDUCCKQSNXPFEGT-NVMNQCDNSA-N
SMILESOC1=CC=CC=C1\C=N/C1=C2C(O)=CC(=CC2=CC(=C1)S(O)(=O)=O)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Proton NMRConsistent with structure
Assay by NMR>95%
Azomethine-H hydrate is used as a highly sensitive and colorimetric reagent for testing and detection of boron. It is also used to detect microgram levels of boron in glass and steel samples. Further, it is involved in the detection of boron in plant samples by masking copper, iron and aluminum ions with ethylenediaminetetraacetic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only