Thermo Scientific Chemicals

Bis(cyclopentadienyl)cobalt, Thermo Scientific Chemicals

Catalog number: 016857.03
1 g, Each
Thermo Scientific Chemicals

Bis(cyclopentadienyl)cobalt, Thermo Scientific Chemicals

Catalog number: 016857.03
1 g, Each
Quantity
Catalog number: 016857.03
also known as 016857-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1277-43-6
IUPAC Name
λ²-cobalt(2+) bis(cyclopenta-2,4-dien-1-ide)
Molecular Formula
C10H10Co
InChI Key
ILZSSCVGGYJLOG-UHFFFAOYSA-N
SMILES
[Co++].[CH-]1C=CC=C1.[CH-]1C=CC=C1
Form
Crystals or crystalline powder and /or chunks
Elemental Analysis
C ≥62.2%
Appearance (Color)
Purple to black

Description

Bis(cyclopentadienyl)cobalt is used to catalyze the preparation of pyridines from alkynes and nitriles. It acts as a polymerization inhibitor of olefins. It is a common one-electron reducing agent in the laboratory. It reacts with carbon monoxide to prepare cobalt(I) derivative, cyclopentadienylcobalt dicarbonyl. It also serves as an internal standard in cyclic voltametry. Further, it finds use in Diels-Alder reactions. In addition to this, it is used as a paint drier and as an oxygen stripping agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text