Thermo Scientific Chemicals

Bismuth(III) oxide, typically 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 017132.30
250 g, Each
Thermo Scientific Chemicals

Bismuth(III) oxide, typically 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 017132.30
250 g, Each
Quantity
Catalog number: 017132.30
also known as 017132-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1304-76-3
IUPAC Name
dibismuth(3+) trioxidandiide
Molecular Formula
Bi2O3
InChI Key
TYIXMATWDRGMPF-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Bi+3].[Bi+3]
Total Metal Impurities
0.01% max.

Description

Used as an adhesive and sealant chemical, flame retardant, pigment, propellant and blowing agent. It is also used as a solvent. Other uses are in batteries, electrical and electronic products, explosive materials, ink, toner, and colorant products, paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text