Thermo Scientific Chemicals

Tris(trimethylsilyl) phosphite, 95%, Thermo Scientific Chemicals

Catalog number: 017159.02
0.5 g, Each
Thermo Scientific Chemicals

Tris(trimethylsilyl) phosphite, 95%, Thermo Scientific Chemicals

Catalog number: 017159.02
0.5 g, Each
Quantity
Catalog number: 017159.02
also known as 017159-02
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1795-31-9
IUPAC Name
tritrimethylsilyl phosphite
Molecular Formula
C9H27O3PSi3
InChI Key
VMZOBROUFBEGAR-UHFFFAOYSA-N
SMILES
C[Si](C)(C)OP(O[Si](C)(C)C)O[Si](C)(C)C
Assay from Supplier's CofA
≥94.0% (titration)
Appearance (Color)
Clear colorless
Form
Liquid

Description

Tris(trimethylsilyl) phosphite is a reagent used for the mild preparation of bis(TMS) phosphonates (Arbuzow reaction). It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text