Thermo Scientific Chemicals

Arsenic(II) sulfide, tech. 90%, Thermo Scientific Chemicals

Catalog number: 017284.06
5 g, Each
Thermo Scientific Chemicals

Arsenic(II) sulfide, tech. 90%, Thermo Scientific Chemicals

Catalog number: 017284.06
5 g, Each
Quantity
Catalog number: 017284.06
also known as 017284-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12044-79-0
IUPAC Name
arsanethiol
Molecular Formula
AsH3S
InChI Key
BMOFEAKQXGGUPE-UHFFFAOYSA-N
SMILES
S[AsH2]
Assay (unspecified)
88.1% min

Description

Arsenic(II) sulfide is used as an primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text