Thermo Scientific Chemicals

n-Pentane, 98%, Thermo Scientific Chemicals

Catalog number: 017381.AK
250 mL, Each
Thermo Scientific Chemicals

n-Pentane, 98%, Thermo Scientific Chemicals

Catalog number: 017381.AK
250 mL, Each
Quantity
Catalog number: 017381.AK
also known as 017381-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-66-0
IUPAC Name
pentane
Molecular Formula
C5H12
InChI Key
OFBQJSOFQDEBGM-UHFFFAOYSA-N
SMILES
CCCCC
Form
Liquid
Water Content (Karl Fischer Titration)
≤0.50%
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%

Description

n-Pentane is used in consumer merchandise such as spot lifters/cleaners and foaming shave gels and in commercial products such as a blowing agent for expanded polystyrene (foam) insulation. It may also be used as a solvent (in pure form) in laboratory, chemical analysis, and other settings where a highly volatile non-polar solvent is needed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text