Zirconium(IV) iodide, 99%, Thermo Scientific Chemicals
Zirconium(IV) iodide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium(IV) iodide, 99%, Thermo Scientific Chemicals

Light, Air and Moisture Sensitive
Have Questions?
Quantity:
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 017400.14
also known as 017400-14
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS13986-26-0
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
Used in the van Arkel-de Boer refining process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only