Iron(II) sulfide, 99% (metals basis), Thermo Scientific Chemicals
Iron(II) sulfide, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iron(II) sulfide, 99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 017422.09
also known as 017422-09
Price (USD)/ Each
53.65
Online exclusive
59.90 
Save 6.25 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
53.65
Online exclusive
59.90 
Save 6.25 (10%)
Add to cart
Iron(II) sulfide, 99% (metals basis), Thermo Scientific Chemicals
Catalog number017422.09
Price (USD)/ Each
53.65
Online exclusive
59.90 
Save 6.25 (10%)
-
Add to cart
Chemical Identifiers
CAS1317-37-9
IUPAC Nameλ²-iron(2+) sulfanediide
Molecular FormulaFeS
InChI KeyGNVXPFBEZCSHQZ-UHFFFAOYSA-N
SMILES[S--].[Fe++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Iron(II) sulfide is used to generate hydrogen sulfide instantaneously by reacting with hydrochloric acid. In laboratory, Kipp’s apparatus is utilized for this purpose. It is used as a re-sulphurizing and alloying agent, as a reducing agent to remove heavy metal impurities from phosphoric acid. It is used to control hydrogen embrittlement in alloy and stainless steel industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only