Vinylferrocene, Thermo Scientific Chemicals
Vinylferrocene, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vinylferrocene, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 017450.03
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1271-51-8
IUPAC NameVinylferrocene
Molecular FormulaC12H12Fe
SMILES[Fe].c1cccc1.C=Cc1cccc1
Molecular Weight (g/mol)212.07
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Appearance (Color)Yellow to brown
Purity≥95.0%
Used in the synthesis of 2-aminomethylferrocenes from vinyl ferrocene and lithium amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only