Thermo Scientific Chemicals

Vinylferrocene, Thermo Scientific Chemicals

Catalog number: 017450.06
5 g, Each
Thermo Scientific Chemicals

Vinylferrocene, Thermo Scientific Chemicals

Catalog number: 017450.06
5 g, Each
Quantity
Catalog number: 017450.06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1271-51-8
IUPAC Name
Vinylferrocene
Molecular Formula
C12H12Fe
SMILES
[Fe].c1cccc1.C=Cc1cccc1
Molecular Weight (g/mol)
212.07
Form
Powder or crystals or crystalline powder
Appearance (Color)
Yellow to brown
Purity
≥95.0%

Description

Used in the synthesis of 2-aminomethylferrocenes from vinyl ferrocene and lithium amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text