Thermo Scientific Chemicals

Tetra-n-propylammonium hydroxide, 40% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 017456.22
100 g, Each
Thermo Scientific Chemicals

Tetra-n-propylammonium hydroxide, 40% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 017456.22
100 g, Each
Quantity
Catalog number: 017456.22
also known as 017456-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4499-86-9
IUPAC Name
tetrapropylazanium hydroxide
Molecular Formula
C12H29NO
InChI Key
LPSKDVINWQNWFE-UHFFFAOYSA-M
SMILES
[OH-].CCC[N+](CCC)(CCC)CCC
Appearance (Color)
Colorless to faintly yellow
Form
Liquid
Assay (unspecified)
Halide: 0.5% max
Elemental Analysis
Sodium + Potassium: 25 ppm, max
Assay (Titration)
38.5 to 40.5%

Description

Tetra-n-propylammonium hydroxide is used as an intermediate, ion exchange agent, surface-active agent and solvent. Further, it is used as an antistatic agent, detergent sanitizers, softener for textiles and paper products, emulsifying agents and pigment dispersers. It finds application as curing accelerators. In addition to this, it acts as a phase transfer catalyst in organic synthesis to enhance the reaction rate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text