Thermo Scientific Chemicals

Iron(III) hydroxide, gamma-phase, Thermo Scientific Chemicals

Catalog number: 017531.18
50 g, Each
Thermo Scientific Chemicals

Iron(III) hydroxide, gamma-phase, Thermo Scientific Chemicals

Catalog number: 017531.18
50 g, Each
Quantity
Catalog number: 017531.18
also known as 017531-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20344-49-4
IUPAC Name
ironoylol
Molecular Formula
FeHO2
InChI Key
AEIXRCIKZIZYPM-UHFFFAOYSA-M
SMILES
O[Fe]=O
X-Ray Diffraction (XRD)
Matches reference

Description

Iron(III) hydroxide, γ-phase is used in purifying water and as an absorbent in chemical processes. It is involved in the preparation of iron oxide-hydroxides nanoparticles, which is used as a very good adsorbent for lead removal from aquatic media. It is also used as a pigment, as a catalyst and in aquarium water treatment as a phosphate binder. Further, it is employed in the paints-lacquers-varnishes industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text