Thermo Scientific Chemicals
Copper(I) phenylacetylide, Thermo Scientific Chemicals
Catalog number: 017538.04
2 g, Each
Thermo Scientific Chemicals
Copper(I) phenylacetylide, Thermo Scientific Chemicals
Catalog number: 017538.04
2 g, Each
Quantity
Catalog number: 017538.04
also known as 017538-04
Price (USD)
Quantity
-
Have Questions?
Description
Copper(I) phenylacetylide participates in efficient coupling with a range of aryl iodides and bromides to give substituted tolanes which, if possessing an ortho amino or hydroxy group, can be thermolyzed to yield 2-phenylbenzofurans, -indoles, and -furo[3,2-b]pyridines.