Lead(II) trifluoroacetate hemihydrate, Thermo Scientific Chemicals
Lead(II) trifluoroacetate hemihydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead(II) trifluoroacetate hemihydrate, Thermo Scientific Chemicals

Have Questions?
Quantity:
10 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 017553.09
also known as 017553-09
Price (USD)/ Each
143.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
169.00 
Save 25.35 (15%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
143.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
169.00 
Save 25.35 (15%)
Add to cart
Lead(II) trifluoroacetate hemihydrate, Thermo Scientific Chemicals
Catalog number017553.09
Price (USD)/ Each
143.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
169.00 
Save 25.35 (15%)
-
Add to cart
Chemical Identifiers
CAS4146-73-0
IUPAC Nameλ²-lead(2+) ditrifluoroacetate
Molecular FormulaC4F6O4Pb
InChI KeyXVULNTYAIWEMSW-UHFFFAOYSA-L
SMILES[Pb++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline solid
Appearance (Color)White to pale cream
Assay from Supplier's CofA≥94%
Melting Point147-153?C
It is employed in pyrolytic deposition of fluoride glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only