Thermo Scientific™

Sodium orthosilicate, (mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution), Thermo Scientific Chemicals

Catalog number: 017568.A1
1 kg, Each
Thermo Scientific™

Sodium orthosilicate, (mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution), Thermo Scientific Chemicals

Catalog number: 017568.A1
1 kg, Each
Quantity
Catalog number: 017568.A1
also known as 017568-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13472-30-5
IUPAC Name
tetrasodium silicate
Molecular Formula
Na4O4Si
InChI Key
POWFTOSLLWLEBN-UHFFFAOYSA-N
SMILES
[Na+].[Na+].[Na+].[Na+].[O-][Si]([O-])([O-])[O-]
Comment
≈ Na_4SiO_4 in solution

Description

It is used as a degreasant, industrial alkaline detergents, water treatment (boiler-feed); adhesives, builder in soaps.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text