Antimony(V) chloride, 99.997% (metals basis), Thermo Scientific Chemicals
Antimony(V) chloride, 99.997% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Antimony(V) chloride, 99.997% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 017570.14
also known as 017570-14
Price (USD)/ Each
161.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
161.00
Add to cart
Antimony(V) chloride, 99.997% (metals basis), Thermo Scientific Chemicals
Catalog number017570.14
Price (USD)/ Each
161.00
-
Add to cart
Chemical Identifiers
CAS7647-18-9
IUPAC Nameantimony pentachloride
Molecular FormulaCl5Sb
InChI KeyVMPVEPPRYRXYNP-UHFFFAOYSA-I
SMILESCl[Sb](Cl)(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.003% max.
Antimony(V) chloride is used as a catalyst and an analytical reagent for testing alkaloids and cesium. It is also used as a polymerization catalyst as well as involved in the chlorination of organic compounds. It acts as a Lewis acid and a strong oxidizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only