Ammonium L-(+)-tartrate, 98%, Thermo Scientific Chemicals
Ammonium L-(+)-tartrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium L-(+)-tartrate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Catalog number 017658.30
also known as 017658-30
Price (USD)/ Each
57.65
Online exclusive
64.40 
Save 6.75 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
57.65
Online exclusive
64.40 
Save 6.75 (10%)
Add to cart
Ammonium L-(+)-tartrate, 98%, Thermo Scientific Chemicals
Catalog number017658.30
Price (USD)/ Each
57.65
Online exclusive
64.40 
Save 6.75 (10%)
-
Add to cart
Chemical Identifiers
CAS3164-29-2
IUPAC Name(2R,3R)-2,3-dihydroxybutanedioic acid diamine
Molecular FormulaC4H12N2O6
InChI KeyNGPGDYLVALNKEG-OLXYHTOASA-N
SMILESN.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Assay (Titration)> 97.5%
Chloride content5 ppm max
CommentPhosphate content: 5 ppm max
CommentSulfate: 0.005% max
View more
Ammonium L-(+)-tartrate finds application in the textile and pharmaceutical industries. It is also used to study protein structural analysis, biochemicals and reagents, biological buffers, chelators and proteomics. It acts as a nitrogen source in biological applications and cell culture media. Further, it is used in the study of crystal optimization research and preliminary diffraction data analysis of the Smad1 MH1 domain bonded to a palindromic SBE DNA element.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only