Thermo Scientific Chemicals

Ammonium L-(+)-tartrate, 98%, Thermo Scientific Chemicals

Catalog number: 017658.A1
1 kg, Each
Thermo Scientific Chemicals

Ammonium L-(+)-tartrate, 98%, Thermo Scientific Chemicals

Catalog number: 017658.A1
1 kg, Each
Quantity
Catalog number: 017658.A1
also known as 017658-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3164-29-2
IUPAC Name
(2R,3R)-2,3-dihydroxybutanedioic acid diamine
Molecular Formula
C4H12N2O6
InChI Key
NGPGDYLVALNKEG-OLXYHTOASA-N
SMILES
N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O
Form
Crystalline powder
Assay (Titration)
> 97.5%
Chloride content
5 ppm max
Comment
Phosphate content: 5 ppm max
Comment
Sulfate: 0.005% max

Description

Ammonium L-(+)-tartrate finds application in the textile and pharmaceutical industries. It is also used to study protein structural analysis, biochemicals and reagents, biological buffers, chelators and proteomics. It acts as a nitrogen source in biological applications and cell culture media. Further, it is used in the study of crystal optimization research and preliminary diffraction data analysis of the Smad1 MH1 domain bonded to a palindromic SBE DNA element.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text