Thermo Scientific Chemicals

Bis(2-ethylhexyl) phosphate, 95%, Thermo Scientific Chemicals

Catalog number: 017723.A3
2 kg, Each
Thermo Scientific Chemicals

Bis(2-ethylhexyl) phosphate, 95%, Thermo Scientific Chemicals

Catalog number: 017723.A3
2 kg, Each
Quantity
Catalog number: 017723.A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
298-07-7
IUPAC Name
bis[(2-ethylhexyl)oxy]phosphinic acid
Molecular Formula
C16H35O4P
InChI Key
SEGLCEQVOFDUPX-UHFFFAOYNA-N
SMILES
CCCCC(CC)COP(O)(=O)OCC(CC)CCCC
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Assay (Titration)
≥94.0 to ≤106.0%
Impurity content
Monoethylhexylphosphoric acid: ≤3%
Refractive Index
1.4400-1.4440 @ 20?C

Description

Extraction of metals.Bis(2-ethylhexyl) phosphate is used as a lubricant additive, corrosion inhibitor and a metal extractant. It is involved in the solvent extraction of uranium salts and rare earth metals. Iron(II) ions are extracted after reduction of Iron(III) ions. Further, it is used as a plasticizer and as a solvent in the synthesis of plastic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text