Thermo Scientific Chemicals
Diphenyldithiophosphonic acid, Thermo Scientific Chemicals
Catalog number: 017758.09
10 g, Each
Thermo Scientific Chemicals
Diphenyldithiophosphonic acid, Thermo Scientific Chemicals
Catalog number: 017758.09
10 g, Each
Quantity
Catalog number: 017758.09
also known as 017758-09
Price (USD)
Quantity
-
Have Questions?
Description
Diphenyldithiophosphonic acid is used in place of phenyl phosphate for acylation of nucleotides.