Thermo Scientific Chemicals

Copper(II) selenide, Black pieces, 99.5% (Metals basis), Thermo Scientific Chemicals

Catalog number: 017901.14
25 g, Each
Thermo Scientific Chemicals

Copper(II) selenide, Black pieces, 99.5% (Metals basis), Thermo Scientific Chemicals

Catalog number: 017901.14
25 g, Each
Quantity
Catalog number: 017901.14
also known as 017901-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1317-41-5
IUPAC Name
copper(2+) selandiide
Molecular Formula
CuSe
InChI Key
QMULOZLYOQCZOH-UHFFFAOYSA-N
SMILES
[Cu++].[Se--]
Total Metal Impurities
0.5% max.

Description

In semiconductors, as catalyst in Kjeldahl digestion.Copper(II) selenide is involved in the preparation of nanocrystals, which shows significant photothermal heating with a photothermal transduction efficiency, thereby it is used for photothermal therapy applications. It is used as a semiconductor and ideal solar control coating as well as finds application in solar cell technology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Form: 6mm and down pieces
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text