Thermo Scientific Chemicals

Ammonium phosphomolybdate hydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 017984.22
100 g, Each
Thermo Scientific Chemicals

Ammonium phosphomolybdate hydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 017984.22
100 g, Each
Quantity
Catalog number: 017984.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
54723-94-3
Impurity content
Ca < 0.02%
Impurity content
SO₄ < 0.025%
Insoluble material
< 0.01%
Trace Metals
(as Pb) < 0.005%
Impurity content
Cl < 0.05%

Description

Ammonium phosphomolybdate hydrate is used to prepare aluminosilicate-supported heteropolyacid catalysts, which finds application in the dehydration of glycerol in the gas phase to get acrolein. It is used as an inorganic ion exchanger involved in the removal of cesium from liquid wastes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text