Ammonium phosphomolybdate hydrate, Reagent Grade, Thermo Scientific Chemicals
Ammonium phosphomolybdate hydrate, Reagent Grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium phosphomolybdate hydrate, Reagent Grade, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 017984.36
also known as 017984-36
Price (USD)/ Each
247.65
Online exclusive
275.00 
Save 27.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
247.65
Online exclusive
275.00 
Save 27.35 (10%)
Add to cart
Ammonium phosphomolybdate hydrate, Reagent Grade, Thermo Scientific Chemicals
Catalog number017984.36
Price (USD)/ Each
247.65
Online exclusive
275.00 
Save 27.35 (10%)
-
Add to cart
Chemical Identifiers
CAS54723-94-3
SpecificationsSpecification SheetSpecification Sheet
Impurity contentCa < 0.02%
Impurity contentSO₄ < 0.025%
Insoluble material< 0.01%
Trace Metals(as Pb) < 0.005%
Impurity contentCl < 0.05%
View more
Ammonium phosphomolybdate hydrate is used to prepare aluminosilicate-supported heteropolyacid catalysts, which finds application in the dehydration of glycerol in the gas phase to get acrolein. It is used as an inorganic ion exchanger involved in the removal of cesium from liquid wastes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only