Zinc citrate dihydrate, Reagent Grade, Zn 31% min, Thermo Scientific Chemicals
Zinc citrate dihydrate, Reagent Grade, Zn 31% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc citrate dihydrate, Reagent Grade, Zn 31% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 018129.30
also known as 018129-30
Price (USD)/ Each
43.65
Online exclusive
48.70 
Save 5.05 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
43.65
Online exclusive
48.70 
Save 5.05 (10%)
Add to cart
Zinc citrate dihydrate, Reagent Grade, Zn 31% min, Thermo Scientific Chemicals
Catalog number018129.30
Price (USD)/ Each
43.65
Online exclusive
48.70 
Save 5.05 (10%)
-
Add to cart
Chemical Identifiers
CAS5990-32-9
IUPAC Nametrizinc(2+) bis(2-hydroxypropane-1,2,3-tricarboxylate) dihydrate
Molecular FormulaC12H14O16Zn3
InChI KeyOXAGUGIXGVHDGD-UHFFFAOYSA-H
SMILESO.O.[Zn++].[Zn++].[Zn++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Zn 31% min
Zinc citrate dihydrate is used in dental care products such as toothpastes, mouthwashes and chewing gums due to its antimicrobial and anti-inflammatory effects. It used in used in food supplements, functional foods and beverages. It is used to formulated in pharmaceuticals, and foods as a zinc supplement. It used as intermediate in organic synthesis, agrochemicals and dyestuff. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only