Thermo Scientific Chemicals

Zinc citrate dihydrate, Reagent Grade, Zn 31% min, Thermo Scientific Chemicals

Catalog number: 018129.A1
1 kg, Each
Thermo Scientific Chemicals

Zinc citrate dihydrate, Reagent Grade, Zn 31% min, Thermo Scientific Chemicals

Catalog number: 018129.A1
1 kg, Each
Quantity
Catalog number: 018129.A1
also known as 018129-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5990-32-9
IUPAC Name
trizinc(2+) bis(2-hydroxypropane-1,2,3-tricarboxylate) dihydrate
Molecular Formula
C12H14O16Zn3
InChI Key
OXAGUGIXGVHDGD-UHFFFAOYSA-H
SMILES
O.O.[Zn++].[Zn++].[Zn++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Assay (unspecified)
Zn 31% min

Description

Zinc citrate dihydrate is used in dental care products such as toothpastes, mouthwashes and chewing gums due to its antimicrobial and anti-inflammatory effects. It used in used in food supplements, functional foods and beverages. It is used to formulated in pharmaceuticals, and foods as a zinc supplement. It used as intermediate in organic synthesis, agrochemicals and dyestuff. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text