Indium Tin eutectic ingot, 99.95% (metals basis), Thermo Scientific Chemicals
Indium Tin eutectic ingot, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Indium Tin eutectic ingot, 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
5x 10 g
Catalog number 018159.09
also known as 018159-09
Price (USD)/ Each
92.20
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
92.20
Add to cart
Indium Tin eutectic ingot, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number018159.09
Price (USD)/ Each
92.20
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialIndium Tin eutectic ingot
Melting Point117°C
Recommended StorageAmbient temperatures
Avg. Mol. Wt. or Mol. Wt. RangeIn:Sn; 52:48 wt%
TSCAYes
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only