Indium Tin eutectic ingot, 99.95% (metals basis)
Indium Tin eutectic ingot, 99.95% (metals basis)
Thermo Scientific Chemicals

Indium Tin eutectic ingot, 99.95% (metals basis)

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
5x 10 g
Catalog number 018159.50
also known as 018159-50
Price (USD)
317.65
Online Exclusive
353.00
Save 35.35 (10%)
Each
Add to cart
Quantity:
5x 10 g
Request bulk or custom format
Price (USD)
317.65
Online Exclusive
353.00
Save 35.35 (10%)
Each
Add to cart
Specifications
Assay Percent Notes(metals basis)
Avg. Mol. Wt. or Mol. Wt. RangeIn:Sn; 52:48 wt%
Chemical Name or MaterialIndium Tin eutectic ingot
MDL NumberMFCD00144477
Melting Point117°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link