Thermo Scientific Chemicals

Gallium Tin alloy, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 018161.18
50 g, Each
Thermo Scientific Chemicals

Gallium Tin alloy, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 018161.18
50 g, Each
Quantity
Catalog number: 018161.18
also known as 018161-18
Price (USD)
Quantity
-
Have Questions?
Appearance (Color)
Silver
Form
Low melting solid
Assay from Supplier's CofA
≥99.99% (metals basis)
Assay from Supplier's CofA
Sn:Ga; 8.0:92.0 wt%

Description

Gallium Tin alloy is used to cool computer chips in place of water.  Used in medical thermometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text