1-Bromopyrene, 95%, Thermo Scientific Chemicals
1-Bromopyrene, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromopyrene, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 018280.03
also known as 018280-03
Price (USD)/ Each
56.90
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
56.90
Add to cart
1-Bromopyrene, 95%, Thermo Scientific Chemicals
Catalog number018280.03
Price (USD)/ Each
56.90
-
Add to cart
Chemical Identifiers
CAS1714-29-0
IUPAC Name1-bromopyrene
Molecular FormulaC16H9Br
InChI KeyHYGLETVERPVXOS-UHFFFAOYSA-N
SMILESBrC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
Formpowder
Assay (GC)> 94.0%
1-Bromopyrene is used in the cleavage of DNA in studies. It is also used in the preparation of DNA base analogs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only