Di-n-butyltin diacetate, 95%, Thermo Scientific Chemicals
Di-n-butyltin diacetate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Di-n-butyltin diacetate, 95%, Thermo Scientific Chemicals

As as stabilizer in chlorinated organics, and as a catalyst for condensation reactions
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 018595.18
also known as 018595-18
Price (USD)/ Each
29.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
35.10 
Save 5.45 (16%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
29.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
35.10 
Save 5.45 (16%)
Add to cart
Di-n-butyltin diacetate, 95%, Thermo Scientific Chemicals
Catalog number018595.18
Price (USD)/ Each
29.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
35.10 
Save 5.45 (16%)
-
Add to cart
Chemical Identifiers
CAS1067-33-0
Molecular FormulaC12H26O4Sn
InChI KeyLDZDQDDROJXUBO-UHFFFAOYSA-N
SMILESCC(O)=O.CC(O)=O.CCCC[SnH2]CCCC
Molecular Weight (g/mol)353.05
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)32.2 to 34.2 wt% total Sn
Appearance (Color)Colorless to pale yellow to pale brown
FormLiquid
Assay (unspecified)33.6 to 35.6 wt% acetic acid
As as stabilizer in chlorinated organics, and as a catalyst for condensation reactions.Di-n-butyltin diacetate is used as a stabilizer for polyvinyl chloride. It acts as a catalyst for the preparation of silicone and urethane foams. Further, it is used as a curing agent for silicone elastomers. In addition to this, it is used as a precursor for thin-film tin(IV) oxide gas sensing materials by laser-assisted chemical vapor deposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only